À propos de cet article
Part of the ZymoPURE™ plasmid kits collection, the ZymoPURE™ plasmid miniprep kit features a spin column-based method for the purification of up to 100 µg of ultra-pure endotoxin-free plasmid DNA in less than 15 minutes.
Fast and reliable purification of up to 100 µg of transfection-grade plasmid DNA using a spin-column
- Innovative ZymoPURE™ technology enables elution of ultra-pure endotoxin-free plasmid DNA in as little as 25 µl
The spin-column design also provides zero buffer retention and a low elution volume. ZymoPURE™ technology uses a modified alkaline lysis method and features novel binding chemistry that yields highly concentrated plasmid DNA (up to 3 µg/µl). In addition, the wash regimen has been optimized to ensure the plasmid DNA is free of endotoxins, salt, protein, and RNA. The result is plasmid DNA suitable for transfection, restriction endonuclease digestion, bacterial transformation, PCR amplification, DNA sequencing, and other sensitive downstream applications. As an added convenience, the ZymoPURE™ plasmid miniprep kit contains colored buffers that permit error-free visualization and identification of complete bacterial cell lysis and neutralization.
Spécifications
- Method / Format:Column (silica-based)
- Prep size:Mini
- Refine target molecule:Plasmid
- Sample type:Bacteria
- Target molecule:Plasmid
- Sample size:≤5 ml
- Description:ZymoPURE™ plasmid miniprep kit
- Yield:≤100 µg