Brilliant III Ultra-Fast SYBR® Green ROX QPCR Master Mix, Agilent Technologies
High performance, ultra-sensitive, SYBR® Green QPCR master mix reagent with ROX concentration for reliable quantification across a wide range of targets and templates.
- Reliable and reproducible data
- Novel fast Taq mutant for qPCR results in under 35 minutes Resistant to common QPCR inhibitors ( whole blood, humic acid, NaCl)
- Enhanced rapid hot start capability saves time and reduces primer-dimer formation
- Convenient pre-blended formulations compatible with any SYBR Green dye
Available with low or high ROX concentrations.
- Application:qPCR dye
Frequently Bought Together