Multiple Affinity Removal Column - Mouse 3, Agilent Technologies
Multiple Affinity Removal Column - Mouse 3, Agilent Technologies
Multiple Affinity Removal Column - Mouse 3, Agilent Technologies
Certificates
Multiple Affinity Removal Column - Mouse 3, Agilent Technologies
Multiple Affinity Removal Column - Mouse 3, Agilent Technologies
1 Options Available Below
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
The Multiple Affinity Removal LC Column – Mouse 3 is designed as a simple way to remove THREE interfering high-abundant proteins from mouse plasma samples.
Removal of these abundant proteins improves the subsequent LC/MS and electrophoretic analysis of the serum sample by effectively expanding the dynamic range of the analysis.
Caution: For research use only.
Product Family Options
Product Information
  • Désignation
    Ø int.
    Longueur
    Availability
    Price
  • Multiple Affinity Removal Column - Ms-3 (Mouse)
    4.60 mm
    50 mm
    Log in for availability
    Log in for availability
    Each
    Each - $3,534.00
    $3,534.00
    Each
       $3,534.00
    Minimum order quantity is 1
    Supplier Number: 5188-5217
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Multiple Affinity Removal Column - Ms-3 (Mouse)
    4.60 mm
    100 mm
    Discontinued
    Supplier Number: -
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.