Airfree® Schlenk Vacuum Manifolds, Inert Gas, Souza, Chemglass
Fournisseur: Chemglass
Certificats
À propos de cet article
Double bank manifolds are modular in design, allowing the user tremendous flexibility in performing experiments.
- Unique design, double bank manifold with the valves sealed in a vertical position
- The main ports are spaced 100 mm center to center
This uniquely designed double bank manifold permits the user an unobstructed view of whether the valve is in the open or closed position, while also exposing the PTFE valves to less outgassing (when the front bank of the manifold is used for inert gas). All connection ports are #15 O-Ring connections that incorporate a greaseless O-Ring seal. Adapters from #15 O-Ring joints to standard taper components, serrated hose connections, or 3/8" Cajon® fittings are available to allow the ports to be used with existing glassware.
For eplacement Chem-Vac™ Chem-Cap®, see (17003-150).
Frequently Bought Together