Ward's® Rota-Rack® Tube Racks
Fournisseur: Heathrow Scientific
Certificats
À propos de cet article
Get organized with this fun, interlocked rack.
- Autoclavable
- Polypropylene
- Two versions to accommodate small and large tubes
Rota-Racks® are suitable for a variety of culture, centrifuge, microcentrifuge, round, flat or conical bottomed tubes.
Frequently Bought Together