Ward's Cool Containers
Ward's Cool Containers
Supplier:   Heathrow Scientific
Ward's Cool Containers
Certificates
Ward's Cool Containers
Ward's Cool Containers
3 Options Available Below
Product Details & Documents
Durable containers for chilling temperature-sensitive samples.
  • Ideal for ice and ice/salt slurries
  • Lid nests under bucket for convenience and to save space
  • Tapered design for easy stacking
  • Designed for low temperature use to –196 °C at 1 bar
Ward's cool containers are perfect for the laboratory or classroom. They are made from polyurethane with metal inserts for stability. The close-fit lid keeps evaporation to a minimum and features a comfortable recessed knob for easy handling. The precise pour spout and large handles allow for excess liquid to be removed easily.
Product Family Options
Informations sur le produit
  • Désignation
    Capacité
    Largeur
    Profondeur
    Hauteur
    Top Diameter
    Availability
    Price
  • Black Bucket
    4 L
    7.1"
    9.2"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $241.10
    $241.10
    Log in pour la disponibilité
    Case of 6
    Case of 6 - $1,446.60
    $1,446.60
    Each
       $241.10
    Case of 6
       $1,446.60
    Supplier Number: HS28714KW
  • Emerald Pan
    1 L
    10.5"
    7.7"
    4.5"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $158.50
    $158.50
    Log in pour la disponibilité
    Case of 6
    Case of 6 - $951.00
    $951.00
    Each
       $158.50
    Case of 6
       $951.00
    Supplier Number: HS28721GW
  • Artic Blue Pan
    4 L
    15"
    10"
    5.5"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $243.30
    $243.30
    Log in pour la disponibilité
    Case of 6
    Case of 6 - $1,459.80
    $1,459.80
    Each
       $243.30
    Case of 6
       $1,459.80
    Supplier Number: HS28724BW