$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Canada
ContactUsLinkComponent
Fittings and Connectors
HPLC Male Nuts Fittings, M6 Thread, MicroSolv
HPLC Male Nuts Fittings, M6 Thread, MicroSolv
10803-846
:
MicroSolv Technology Corp
HPLC Male Nuts Fittings, M6 Thread, MicroSolv
10803-846
:
MicroSolv Technology Corp
:
47301-M6
Description:
Male nuts fitttings
Length:
15 mm
Material:
PEEK
Cat. no.:
10803-846
For Use With:
¹/₁₆" tubing
Supplier No.:
47301-M6