Injection Training Pads | Avantor
Injection Training Pads
Supplier:   WALLCUR, LLC.
Injection Training Pads
Supplier:  WALLCUR, LLC.
Certificates
Injection Training Pads
Injection Training Pads
2 Options Available Below
Product Details & Documents
Remedy's medical training injection pads are designed for the practice of needle injections.
The pads can be used to simulate intradermal, intra-muscular and subcutaneous injections. The larger pad allows you to have more practice area, as well as practicing Z-track injections. A removable epidermis layer is placed on the surface of the pad allowing for the simulation of a "wheal" during intradermal injections.
The skin puncture resistance, as well as the resistance throughout the injection, closely mimics the feel of a real injection. Both pads have drainage on the bottom so they can be used with fluid filled syringes.
Ordering information: Additional epidermis layers may be purchased separately.
Product Family Options
Informations sur le produit
  • Désignation
    Width
    Longueur
    Hauteur
    Availability
    Price
  • Small Injection Pad "Paddy"
    2.5"
    2.5"
    1.75"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $57.19
    $57.19
    Each
       $57.19
    Supplier Number: REM-INJ-SP-01L
  • Large Injection Pad "Patrick"
    6"
    6"
    1.75"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $229.29
    $229.29
    Each
       $229.29
    Supplier Number: REM-INJ-LG-01L
  • Epi-Layers
    Discontinued
    Supplier Number: REM-INJ-EPI-01