Commander⢠Safety Glasses, Protective Industrial Products
Supplier: PIP
Certificates
About this item
Lightweight and semi-rimless design offers eye coverage and comfort.
- Bayonet-style temples with non-slip rubber pads provide a secure fit
- Adjustable rubber nose pads
- FogLess⢠lens coating as standard
The 9.5 base curve lens are used with anti fog and anti scratch coating.
Certifications: Meets ANSI Z87.1 and CSA Z94.3 requirements.
Frequently Bought Together