Triangles | Avantor
Triangles
Triangles
Certificates
Triangles
2 Options Available Below
Product Details & Documents
These triangles are made up of iron wire (16 gauge) galvanized with plain clay pipe stems.
Smooth pipe stem triangles protect glassware on hotplates and over burner flames.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Triangles, iron wire, 5.1 cm (2")
    Discontinued
    Supplier Number: -
  • Triangles, iron wire, 6.4 cm (2.5")
    Discontinued
    Supplier Number: H-23870
  • Triangles, iron wire, 5.1 cm (2")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $6.32
    $6.32
    Each
       $6.32
    Supplier Number: H-23860
  • Triangles, iron wire, 6.4 cm (2.5")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4.21
    $4.21
    Each
       $4.21
    Supplier Number: H-23870