Antistatic Polystyrene Hexagonal Weighing Dishes, Simport Scientific
Fournisseur: Simport Scientific
Certificats
À propos de cet article
Ideal for handling solids or liquids during weighing.
- Easily bent into pouring spouts
- Dishes enable non-spill transfer
- Stackable for easy storage
- Withstands temperatures up to 80 °C
Molded hexagonal design provides greater balance protection and safety. Will not react with most substances. Suitable for weighing static-affected samples.
Frequently Bought Together