Dissecting Forcep, Walter Stern
Dissecting forceps, 114 mm (4½"), Curved
Catalog # 25607-196
Supplier: Walter Stern
Supplier #: 300-131
Jump to:
Product Details & Documents
This stainless steel dissecting forcep features fine point, serrated tips and guide pins.
Specifications
- Désignation:Dissecting forceps
- Longueur:114 mm (4½")
- Tip Style:Curved