ContactUsLinkComponent
STACCUPS® Graduated Beakers, PP, Cargille
  13984-004
 : 6619
STACCUPS® Graduated Beakers, PP, Cargille
  13984-004
 :  06604
 : 6619
Select an Option
25 ml
Select an Option
25 - 500 ml
Select an Option
500 ml
Select an Option
13984-004
Select an Option
6619
STACCUPS® sterilizable graduated beakers are made from heavy-gauge PP and reusable.
  • Chemically unreactive to most solvents, acids, alkaline, salts, organics and biologicals
  • Seamless one-piece construction makes STACCUPS® leak-proof
  • Pouring spout (360°) - gentle squeeze creates a controllable pouring spout on the STACCUPS'® circumference - adaptable for any rate of decanting
Resistant to temperatures of up to 121 °C (250 °F) and also can be subjected to very low temperatures for sample deep-freeze and storage.
STACCUPS® are stack-packed with a protective shrink-wrap which doubles as a self-dispenser and allows for cleanliness and convenient storing.
Unique, underside rim provides maximum stability and special rib prevents STACCUPS® from sticking when nested - helps save storage space.
Frequently Bought Together