ContactUsLinkComponent
MVP Human Total RNA, Agilent Technologies
Catalog # CA99901-924
CAS Number:  
undefined
MVP Human Total RNA, Agilent Technologies
Catalog # CA99901-924
Supplier Number:  540025
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Total RNA, Placenta, Human
  • Size:
    25 µg
  • Cat. no.:
    CA99901-924
Specifications
About this item
MVP RNA is application-ready RNA available in small economical pack sizes.
  • Eliminates tedious, time consuming RNA isolation procedures
  • Small, economically priced and convenient 25 µg pack sizes
  • Matched sets of normal and diseased tissue from the same donor
  • Well-documented donor, tissue pathology information
Extensive and rigorous quality control including many application specific assays provides assurance that the RNA is intact, full-length and nearly DNA-free.
Application: Real time RT-PCR, miRNA detection and Northern blot analysis.
Caution: For research use only. Not for use in diagnostic procedures.