dPette+ Electronic Pipettes, Variable Volume, DLAB Scientific | Avantor
dPette+ Electronic Pipettes, Variable Volume, DLAB Scientific
dPette+ Electronic Pipettes, Variable Volume, DLAB Scientific
Certificates
dPette+ Electronic Pipettes, Variable Volume, DLAB Scientific
dPette+ Electronic Pipettes, Variable Volume, DLAB Scientific
4 Options Available Below
Product Details & Documents
High accuracy, high performance stepper motor ensures accuracy and repeatability, eliminating manual pipetting errors.
  • Two buttons handle all operational settings
  • Lightweight, ergonomic design
  • Small body dimensions enable easy handling and fatigue-free pipetting
  • Adjustable speeds for aspiration and dispensing
  • Li-ion battery and dual charging modes enable longer operation time
  • Self calibration via USB to PC; third party calibration is not required
A motor driven digital control pipette with multifunctional capabilities of pipetting, mixing, stepper, and dilution.
Electrical: 100 to 220 V, 50/60 Hz
 : CE marked. cTUVus.
 : Each package includes the electronic pipette with battery, stand, charger, USB cable, calibration card, and user manual.
Product Family Options
Informations sur le produit
  • Désignation
    Volume
    Increments
    Availability
    Price
  • dPette+ Electronic Pipette, with US Plug
    0.5 - 10 µl
    0.01 µl
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,007.13
    $1,007.13
    Each
       $1,007.13
    Supplier Number: 7016201001
  • dPette+ Electronic Pipette, with US Plug
    5 - 50 µl
    0.1 µl
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,007.13
    $1,007.13
    Each
       $1,007.13
    La quantité minimale de commande est de 1
    Supplier Number: 7016201002
  • dPette+ Electronic Pipette, with US Plug
    30 - 300 µl
    1 µl
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $967.10
    $967.10
    Each
       $967.10
    Supplier Number: 7016201003
  • dPette+ Electronic Pipette, with US Plug
    100 - 1000 µl
    5 µl
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,007.13
    $1,007.13
    Each
       $1,007.13
    Supplier Number: 7016201004