Sling Psychrometer | Avantor
Sling Psychrometer
Sling Psychrometer
Certificates
Sling Psychrometer
Sling Psychrometer
2 Options Available Below
Product Details & Documents
Use this simple apparatus to measure relative humidity.
  • Requires assembly
The thermometers mount on a wooden handle so they can be spun to obtain temperature readings.
Ordering information: Includes an activity guide with relative humidity calculation table. Includes two plastic-backed Celsius thermometers, one for dry bulb measurements and the other for wet bulb measurements.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Sling Psychrometer
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $23.85
    $23.85
    Each
       $23.85
    Supplier Number: SPCRKIT
  • Sling Psychrometer, Pack of 15
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 15
    Pack of 15 - $165.90
    $165.90
    Pack of 15
       $165.90
    Supplier Number: SPCRKIT-PK/15