CENCO® Hall Effect
CENCO® Hall Effect Apparatus
  470303-942
 :  HLEF01-VWR
CENCO® Hall Effect
  470303-942
CENCO® Hall Effect
CENCO® Hall Effect
CENCO® Hall Effect Apparatus
$4,981.40
Each
Explore the Hall Effect in a GaAs semiconductor.
  • n-type GaAs Hall sensor chip
  • Helmholtz coil pair 75 mm effective diameter, 11.25 mT central magnetic induction at 0.5 A
  • Two-axis mechanical traverse for field plotting
  • Constant current sources for magnet (0 to 0.5 A) and Hall driving current (0 to 3 mA)
  • Reversing switch for exploring the elimination of systematic error
  • 31/2 digit current and voltage meters
  • Metal storage case for the Hall Effect board
Explores systematic errors and their elimination. Determines the conductivity of the semiconductor.
This Hall Effect Apparatus consists of a mounted n-type semiconductor chip on a traverse mechanism, a pair of coils, relay-controlled reversing switches, and a control unit with connecting cords.
Uses the semiconductor to plot the coils magnetic field.
The Hall Effect is the generation of a side-to-side voltage in a conductor or semiconductor carrying a current when it is placed in a magnetic field.
The effect is widely used in magnetic field sensors. It is associated with several well-known systematic errors, some of which can be eliminated by special measurement techniques.
  • Désignation:
    CENCO® Hall Effect Apparatus