Brilliant III Probe MM, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
Certificates
About this item
Brilliant III QPCR and QRT-PCR master mixes increase overall sensitivity and speed through the novel, quick-activating hot start and the engineered Taq-mutant.
- Extremely fast while maintaining sensitivity
- Greater resistance to common QPCR inhibitors (whole blood or NaCl)
- Optimized fast cycling formulation ensures reliable and reproducible data with shorter run times
- Convenient pre-blended formulations compatible with any sequence-specific probe detection chemistry
- Designed for researchers who want access to their data faster without compromising data quality
Brilliant III Ultra-Fast maintains amplification efficiency, R2, dynamic range and detection sensitivity all while shortening the total cycling time by approximately 60% compared to standard cycling.
Caution: For research use only. Not for use in diagnostic procedures.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Application:qPCR probe