ContactUsLinkComponent
Rotavapor® R-220 Pro Rotary Evaporators, 230 V
 :  Buchi
undefined
Rotavapor® R-220 Pro Rotary Evaporators, 230 V
 :  Buchi
  • Catalog No:
  • 76266-302
  • 76266-276
  • 76266-298
  • 76266-300
  • Model:
  • R-220 Pro Essential
  • R-220 Pro
  • R-220 Pro
  • R-220 Pro
  • Rotation speed:
  • 5 - 150 rpm
  • 5 - 150 rpm
  • 5 - 150 rpm
  • 5 - 150 rpm
  • Weight:
  • 85 kg
  • 100 kg
  • 100 kg
  • 100 kg
  • Dimensions:
  • 1100W×560D×1700H mm
  • 1200W×710D×1750H mm
  • 1200W×710D×1750H mm
  • 1200W×710D×1750H mm
Frequently Bought Together