Vernier® Go Direct™ Chloride Ion-Selective Electrode | Avantor
Vernier® Go Direct™ Chloride Ion-Selective Electrode
Go Direct™ Chloride ion-selective electrode
Catalog # CA470303-902
New Product
Fournisseur #:  GDX-CL
Vernier® Go Direct™ Chloride Ion-Selective Electrode
Catalog # CA470303-902
New Product
Vernier® Go Direct™ Chloride Ion-Selective Electrode
Vernier® Go Direct™ Chloride Ion-Selective Electrode
Go Direct™ Chloride ion-selective electrode
Price
$537.09
Log in pour la disponibilité
Each
La quantité minimale de commande est de 1
Product Details & Documents
The Go Direct Chloride Ion-Selective Electrode (ISE) is used to measure the concentration of chloride (Cl-) in aqueous samples.
The concentration of chloride gives a quick measurement of the salinity of water samples. Use the Go Direct Chloride Ion-Selective Electrode to wirelessly measure chloride levels in ocean saltwater or salt in food samples.
The versatile BNC connector allows students to easily replace the electrode when it expires or becomes damaged. You need only replace the electrode and not the entire sensor-lowering your overall equipment costs.
The Go Direct Chloride Ion-Selective Electrode has a combination-style, non-refillable, gel-filled electrode. Unlike other ISEs available from Vernier, the Go Direct Chloride Ion-Selective Electrode has a solid-state membrane that does not need to be replaced.
The Go Direct robust rechargeable battery offers long battery life and provides always-ready operation. Monitor battery life directly from our free Graphical Analysis™ 4 app. Should the battery run low, simply connect the sensor to the charging cable and keep on collecting-no interruptions or inconvenience to you or your students.
Our Go Direct sensors connect directly to your mobile device, Chromebook™, or computer using our free Graphical Analysis™ 4 app-no additional equipment or software purchases are necessary. Go Direct sensors can be used wired via USB or wirelessly via Bluetooth wireless technology, allowing you to choose the solution best for your classroom or laboratory.
 : Important: Ion-Selective Electrodes require good chemical technique and careful calibration to obtain accurate results; they are not recommended for middle school or elementary students.
Spécifications
  • Désignation:
    Go Direct™ Chloride ion-selective electrode
Accessories
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Chloride standard solution, high
    Chloride standard solution, high
    CA470359-906
    Log in pour la disponibilité
    Each - $45.09
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Chloride standard solution, low
    Chloride standard solution, low
    CA470359-908
    Log in pour la disponibilité
    Each - $45.09