Synthware Single Volume Pipettes, Kemtech America
Supplier: Kemtech America
Certificates
About this item
These single volume pipettes are certified class A.
- High quality borosilicate glass
Synthware single volume pipets are perfect for transferring small amounts of liquid.
Certifications: Meets ASTM Specification E438, Type 1, Class A, and 'Standard Specification for Glasses in Laboratory Apparatus'.
Frequently Bought Together