Laboratory Trays, United Scientific Supplies
Supplier: United Scientific Supplies
Certificates
About this item
General use tray for all laboratory needs.
- Polypropylene
- Autoclavable
Multipurpose, polypropylene molded trays can be used for sterilizing, drying glassware, porcelain ware, and other laboratory supplies. These steam autoclavable trays have tapered walls designed for nesting.
Frequently Bought Together