Disposable Cuvettes for Fluorometer, FireflySci
Disposable Cuvettes for Fluorometer, FireflySci
Supplier:   FIREFLYSCI, INC.
Disposable Cuvettes for Fluorometer, FireflySci
Supplier:  FIREFLYSCI, INC.
Certificates
Disposable Cuvettes for Fluorometer, FireflySci
Disposable Cuvettes for Fluorometer, FireflySci
3 Options Available Below
Product Details & Documents
These cuvettes are used for rapid assays and comes in different materials polystyrene and acrylic.
  • Four windows and base optically polished
  • With open top
Product Family Options
Informations sur le produit
  • Désignation
    Volume
    Internal Dimensions
    Outer Depth
    Outer Height
    Outer Width
    Lightpath
    Matière
    Availability
    Price
  • Disposable cuvette, type 9FLP, semi-micro
    1.500 µl
    L×W 10×4 mm
    12.5 mm
    45 mm
    12.5 mm
    10 mm
    Polystyrene
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $443.24
    $443.24
    Each
       $443.24
    Supplier Number: 9FLPMMA
  • Disposable cuvette, type 1FLP, macro
    3.500 µl
    L×W 10×10 mm
    12.5 mm
    45 mm
    12.5 mm
    10 mm
    Polystyrene
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $232.95
    $232.95
    Prix promotionnel pour les nouveaux clients
    Each
       $232.95
    Supplier Number: 1FLPS
  • Disposable cuvette, type 1FLP, macro
    3.500 µl
    L×W 10×10 mm
    12.5 mm
    45 mm
    12.5 mm
    10 mm
    Acrylic
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $369.72
    $369.72
    Each
       $369.72
    Supplier Number: 1FLPMMA