ContactUsLinkComponent
Printer Beakers, United Scientific Supplies
  76263-410
 : P50606
undefined
Printer Beakers, United Scientific Supplies
  76263-410
 :  P50601
 : P50606
Select an Option
155 mm
Select an Option
100 ml
Select an Option
200 - 2000 ml
Select an Option
2000 ml
Select an Option
190 mm
Select an Option
76263-410
Perfectly sized for general labware use.
  • Tapered design is convenient for stacking and holding
  • All sizes have easy to read silk-screened graduations
  • Autoclavable
Polypropylene beakers with pouring spouts are ideal for general laboratory use.
Frequently Bought Together