Laboratory Scoops, United Scientific Supplies
Fournisseur: United Scientific Supplies
Certificats
À propos de cet article
Conveniently designed handled scoops.
- Autoclavable
These sturdy polypropylene scoops are available in a variety of sizes, as well as in a complete set of seven scoops. Capacities are molded into the top of each scoop for easy reference. The polypropylene material is autoclavable and will not rust or corrode.
Frequently Bought Together