Acrylic Burettes, United Scientific Supplies
These hardy burettes are designed for general use.
- Acrylic
- Transparent
- Leakproof stopcock included
- Unaffected by most dilute aqueous solutions
- Heavy-duty construction
Clear acrylic body and leakproof stopcock with a self lubricating PTFE plug. The acrylic body is resistant to dilute mineral acids and bases, except Hydrofluoric Acid and Ammonium Hydroxide. Graduations are in 0.1 ml increments.
Frequently Bought Together