About this item
Made of acetal and resistant to the chemical action of most histological solvents and suitable for automated printers.
- In Quickload™ stacks, for Primera printers
- 35° anterior writing area
- One-piece construction with hinge-lock design prevents early separation of base and lid
- Snap latch and hinge-lock design; cassettes will always relock securely
- Allows for one-hand operation
Secure specimens during processing with the Unisette™ Biopsy Processing/Embedding Cassette. The snap latch and hinge-lock design allows for one-hand operation and always relocks securely without danger of specimen loss.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.