Slimsette™ Biopsy Processing/Embedding Cassette in Quickload™ Stacks, Simport Scientific
À propos de cet article
Secure specimens during processing with the Slimsette™ Biopsy Processing/Embedding Cassette, in Quickload™ stacks.
- In Quickload™ stacks
- 45° anterior writing area
- Recessed cover
- Suitable for automated printers
- Made of acetal
- Resistant to the chemical action of most histological solvent
Certifications: CE-IVD.
Packaging: Each case contains 50 Quickload™ stacks of 40 cassettes.
11 Options Available Below
Product Details & Documents
Secure specimens during processing with the Slimsette™ Biopsy Processing/Embedding Cassette, in Quickload™ stacks.
- In Quickload™ stacks
- 45° anterior writing area
- Recessed cover
- Suitable for automated printers
- Made of acetal
- Resistant to the chemical action of most histological solvent
Certifications: CE-IVD.
Packaging: Each case contains 50 Quickload™ stacks of 40 cassettes.
Product Family Options
Informations sur le produit
- ColorAvailabilityPrice
- Supplier Number: M510-12T
- Supplier Number: M510-6T
- Supplier Number: M510-9T
- Supplier Number: M510-4T
- Supplier Number: M510-10T
- Supplier Number: M510-11T
- Supplier Number: M510-7T
- Supplier Number: M510-3T
- Supplier Number: M510-8T
- Supplier Number: M510-2T
- Supplier Number: M510-5T