Unisette™ Tissue Processing/Embedding Cassette in Quickload™ Stacks, Simport
Unisette™ Tissue Processing/Embedding Cassette in Quickload™ Stacks, Simport
Unisette™ Tissue Processing/Embedding Cassette in Quickload™ Stacks, Simport
Supplier:  Simport Scientific
Certificates
Unisette™ Tissue Processing/Embedding Cassette in Quickload™ Stacks, Simport
Unisette™ Tissue Processing/Embedding Cassette in Quickload™ Stacks, Simport
11 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Secure specimens during processing with the Unisette™ Tissue Processing/Embedding Cassette.
  • In Quickload™ stacks
  • 35° Anterior writing area
  • One-piece construction with hinge-lock design prevents early separation of base and lid
  • Snap latch and hinge-lock design; cassettes will always relock securely
  • Allows for one-hand operation
  • Suitable for automated printers
  • Made of acetal
  • Resistant to the chemical action of most histological solvents
The snap latch and hinge-lock design allows for one-hand operation and always relocks securely without danger of specimen loss.
Certifications: CE-IVD.
Packaging: Each case contains 25 Quickload™ stacks of 40 cassettes.
Product Family Options
Product Information
  • Color
    Availability
    Price
  • Aqua
    Log in for availability
    Log in for availability
    Case of 1,000
    Case of 1,000 - $394.35
    $394.35
    Case of 1,000
       $394.35
    Supplier Number: M505-12T
  • Blue
    Log in for availability
    Log in for availability
    Case of 1,000
    Case of 1,000 - $444.00
    $444.00
    Case of 1,000
       $444.00
    Supplier Number: M505-6T
  • Gray
    Log in for availability
    Log in for availability
    Case of 1,000
    Case of 1,000 - $443.16
    $443.16
    Case of 1,000
       $443.16
    Supplier Number: M505-9T
  • Green
    Log in for availability
    Log in for availability
    Case of 1,000
    Case of 1,000 - $444.00
    $444.00
    Case of 1,000
       $444.00
    Supplier Number: M505-4T
  • Lilac
    Log in for availability
    Log in for availability
    Case of 1,000
    Case of 1,000 - $394.35
    $394.35
    Case of 1,000
       $394.35
    Supplier Number: M505-10T
  • Orange
    Log in for availability
    Log in for availability
    Case of 1,000
    Case of 1,000 - $394.35
    $394.35
    Case of 1,000
       $394.35
    Supplier Number: M505-11T
  • Peach
    Log in for availability
    Log in for availability
    Case of 1,000
    Case of 1,000 - $394.35
    $394.35
    Case of 1,000
       $394.35
    Supplier Number: M505-7T
  • Pink
    Log in for availability
    Log in for availability
    Case of 1,000
    Case of 1,000 - $444.00
    $444.00
    Case of 1,000
       $444.00
    Supplier Number: M505-3T
  • Tan
    Log in for availability
    Log in for availability
    Case of 1,000
    Case of 1,000 - $444.00
    $444.00
    Case of 1,000
       $444.00
    Supplier Number: M505-8T
  • White
    Log in for availability
    Log in for availability
    Case of 1,000
    Case of 1,000 - $444.00
    $444.00
    Case of 1,000
       $444.00
    Supplier Number: M505-2T
  • Yellow
    Log in for availability
    Log in for availability
    Case of 1,000
    Case of 1,000 - $524.53
    $524.53
    Case of 1,000
       $524.53
    Supplier Number: M505-5T