Micro Fluorescence Cuvettes with PTFE Stopper, Type 30FL, FireflySci
Fournisseur: FIREFLYSCI, INC.
Certificats
À propos de cet article
The Type 30FL cuvettes are made with optical glass, UV quartz, IR quartz with transmission values.
- Fire-fused to ensure that cuvettes are robust and durable
- Four windows and base optically polished
- Highly accurate results, only need to test sample once
- Base thickness: 3 mm
Each cell goes through a rigorous inspection process before leaving our factory. Made by hand, ensuring high quality optical polishing with no defects.
Delivery: Includes PTFE stopper.
Frequently Bought Together