M480 Disposable Embedding Cassettes in Quickload™ Stacks, Simport
Supplier: Simport Scientific
Certificates
About this item
Designed for holding and identifying tissue samples in processing/embedding procedures.
- In Quickload™ stacks
- 45° anterior writing area
- Accept standard metal lids (87002-382)
- Suitable for automated printers
- Made of acetal
- Resistant to the chemical action of most histological solvents
Certifications: CE-IVD.
Packaging: Each case contains 50 Quickload™ stacks of 40 cassettes.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Frequently Bought Together