Swingsette™ Tissue Processing/Embedding Cassette in Quickload™ Stacks, Simport
À propos de cet article
Secure specimens during processing with the Swingsette™ Tissue Processing/Embedding Cassette, in Quickload™ stacks.
- In Quickload™ stacks
- 45° anterior writing area
- Easy to assemble hinged lids
- Suitable for automated printers
- Made of acetal
- Resistant to the chemical action of most histological solvents
Certifications: CE-IVD.
Packaging: Each case contains 50 Quickload™ stacks of 40 cassettes, lids packaged separately.
11 Options Available Below
Product Details & Documents
Secure specimens during processing with the Swingsette™ Tissue Processing/Embedding Cassette, in Quickload™ stacks.
- In Quickload™ stacks
- 45° anterior writing area
- Easy to assemble hinged lids
- Suitable for automated printers
- Made of acetal
- Resistant to the chemical action of most histological solvents
Certifications: CE-IVD.
Packaging: Each case contains 50 Quickload™ stacks of 40 cassettes, lids packaged separately.
Product Family Options
Informations sur le produit
- ColorAvailabilityPrice
- Supplier Number: M517-12T
- Supplier Number: M517-6T
- Supplier Number: M517-9T
- Supplier Number: M517-4T
- Supplier Number: M517-10T
- Supplier Number: M517-11T
- Supplier Number: M517-7T
- Supplier Number: M517-3T
- Supplier Number: M517-8T
- Supplier Number: M517-2T
- Supplier Number: M517-5T