VWR® Turbidity Meter
VWR® Turbidity Meter
76152-030
: 76152-030
Autoranging from 0 to 1000 NTUs, the meter generates an accurate turbidity reading in seconds.
- Infrared LED measurement
- No maintenance
- IP 67 (waterproof) rated
- EPA- approved standards
- Three years warranty
Generate an accurate turbidity reading quickly with the VWR® Turbidity Meter. Perfect for the plant, field or lab, this simple yet powerful turbidity meter offers an impressive array of features. Use the Infrared (IR) LED light source to comply with ISO 7027 requirements and reliably measure turbidity with auto-ranging values from 0 to 1000 NTU. This meter offers a long battery life averaging about 3000 tests.
: TUV 3-in-1, ISO 7027.
: Kit includes batteries, vials, turbidity standards, silicone oil and cloth, and hard sided carrying case.
- Désignation:VWR® Turbidity Meter
- Measurement range:0 – 1000 NTU
- Cat. No.:76152-030
- Sample requirements - temperature:0...50 °C (32...122 °F)
- Precision:±2% of reading ±1 LSD for 0 – 500 NTU;
±3% of reading ±1 LSD for 501 – 1000 NTU - Automatic range selection:0.01 – 19.99 NTU;
20.0 – 99.9 NTU;
100 – 1000 NTU - Resolution:0.01 NTU (0 – 19.99 NTU);
0.1 NTU (20 – 99.9 NTU);
1 NTU (100 – 1000 NTU) - Operating temperature:0...50 °C (32...122 °F)