Fine Mesh Basket, Sklar®
À propos de cet article
All Sklar products are industry standard.
- For more than 120 years, Sklar has set the standard for surgical instrumentation
- With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
Sklar's® Fine Mesh Basket is designed for use with cleaning, sterilizing, and handling small instruments. The baskets are available in a variety of sizes, with or without removable hinges and with or without locks.
22 Options Available Below
Product Details & Documents
All Sklar products are industry standard.
- For more than 120 years, Sklar has set the standard for surgical instrumentation
- With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
Sklar's® Fine Mesh Basket is designed for use with cleaning, sterilizing, and handling small instruments. The baskets are available in a variety of sizes, with or without removable hinges and with or without locks.
Product Family Options
Informations sur le produit
- DésignationMatièreDimensionsAvailabilityPrice
- Supplier Number: 10-6600
- Supplier Number: 10-6602
- Supplier Number: 10-6606
- Supplier Number: 10-6608
- Fine Mesh BasketStainless steel15×103/4×11/2"Log in pour la disponibilitéLog in pour la disponibilitéEachEach - $1,410.84$1,410.84Prix promotionnel pour les nouveaux clientsEach$1,410.84Supplier Number: 10-6604
- Fine Mesh Basket with Locking ClipStainless steel3×11/2×3/4"Log in pour la disponibilitéEach$388.54Supplier Number: 10-6612
- Supplier Number: 10-6614
- Supplier Number: 10-6616
- Supplier Number: 10-6618
- Supplier Number: 10-6620
- Supplier Number: 10-6622
- Supplier Number: 10-6624
- Supplier Number: 10-6626
- Fine Mesh Basket with Locking ClipStainless steel83/4×51/2×2"Log in pour la disponibilitéEach$975.50Supplier Number: 10-6628
- Fine Mesh Basket with Locking ClipStainless steel91/4×6×11/2"Log in pour la disponibilitéEach$942.86Supplier Number: 10-6630
- Supplier Number: 10-6610
- Fine Mesh Basket with Locking ClipStainless steel91/2×31/2×11/2"Log in pour la disponibilitéEach$975.50Supplier Number: 10-6632
- Fine Mesh Basket with Locking ClipStainless steel91/2×5×11/2"Log in pour la disponibilitéEach$942.86Supplier Number: 10-6634
- Fine Mesh Basket with Locking ClipStainless steel11×7×11/2"Log in pour la disponibilitéEach$989.38Supplier Number: 10-6636
- Fine Mesh Basket with Silicone PadsStainless steel/silicone91/4×3×1"Log in pour la disponibilitéEach$942.86Supplier Number: 10-6729
- Fine Mesh Basket with Silicone PadsStainless steel/silicone91/4×6×11/2"Log in pour la disponibilitéEach$942.86Supplier Number: 10-6730
- Fine Mesh Basket with Silicone PadsStainless steel/silicone91/2×31/2×11/2"Log in pour la disponibilitéEach$942.86Supplier Number: 10-6732