Ramelet Phlebectomy Hook, Sklar®
Ramelet Phlebectomy Hook, Sklar®
Supplier:   Sklar
Ramelet Phlebectomy Hook, Sklar®
Supplier:  Sklar
Certificates
Ramelet Phlebectomy Hook, Sklar®
Ramelet Phlebectomy Hook, Sklar®
6 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Sklar's® Ramelet Phlebectomy Hook is used to grip the vein during surgical procedures.
  • With industrial standard
  • With five major surgical instruments line
  • Delivers superior precision for any surgical procedure
The hook comes in fine and medium tips, angled left and right. The handle is round and knurled, which makes it easier to roll between the fingers for better control.
Product Family Options
Product Information
  • Désignation
    Longueur
    Matière
    Curvature
    Availability
    Price
  • Ramelet Phlebectomy Hook, Fine
    10.2 cm (4")
    Stainless steel
    Angled (Left)
    Log in for availability
    Log in for availability
    Each
    Each - $774.37
    $774.37
    New Customer Promotional Price
    Each
       $774.37
    Supplier Number: 06-4173
  • Ramelet Phlebectomy Hook, Medium
    10.2 cm (4")
    Stainless steel
    Angled (Left)
    Log in for availability
    Log in for availability
    Each
    Each - $774.37
    $774.37
    New Customer Promotional Price
    Each
       $774.37
    Supplier Number: 06-4174
  • Ramelet Phlebectomy Hook, Set of Two, 1-Fine, 1- Medium
    10.2 cm (4")
    Stainless steel
    Angled (Left)
    Log in for availability
    Log in for availability
    Each
    Each - $1,548.62
    $1,548.62
    New Customer Promotional Price
    Each
       $1,548.62
    Supplier Number: 06-4175
  • Ramelet Phlebectomy Hook, Fine
    10.2 cm (4")
    Stainless steel
    Angled (Right)
    Log in for availability
    Log in for availability
    Each
    Each - $774.37
    $774.37
    New Customer Promotional Price
    Each
       $774.37
    Supplier Number: 06-4170
  • Ramelet Phlebectomy Hook, Medium
    10.2 cm (4")
    Stainless steel
    Angled (Right)
    Log in for availability
    Log in for availability
    Each
    Each - $774.37
    $774.37
    New Customer Promotional Price
    Each
       $774.37
    Supplier Number: 06-4171
  • Ramelet Phlebectomy Hook, Set of Two, 1-Fine, 1- Medium
    10.2 cm (4")
    Stainless steel
    Angled (Right)
    Log in for availability
    Log in for availability
    Each
    Each - $1,548.62
    $1,548.62
    New Customer Promotional Price
    Each
       $1,548.62
    Supplier Number: 06-4172