Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Spécifications
- Description:Katsch Comedone Extractor
- Length:5½"
- Material:Stainless steel
- Cat. no.:76191-800
- Tip Style:Sharp/Blunt
Spécifications
À propos de cet article
Sklar's® Katsch Comedone Extractor is a dermal instrument used to treat facial blemishes.
- All Sklar products are industry standard
This double-ended instrument has one fenestrated, spoon-shaped end specifically designed for extraction and one spear blade used for scraping. With gentle pressure the instrument assists in forcing the blemish causing debris out from under your skin.