Traube Percussion Hammer, OR-Grade, Sklar®
Catalogue # 76191-946
Fournisseur: Sklar
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
- Description:Traube
- Length:6¼"
- Material:Stainless steel
- Cat. no.:76191-946
- Weight:5 oz.
Spécifications
À propos de cet article
Sklar's® Traube Percussion Hammer is primarily used to check pressure and reflex in the stomach area.
- Single-ended instrument
- Industry standard
- Replacable bumper tip
By using this instrument and a percussion method of examination, the physician may be able to detect early indicators of enlarged organs, which may be caused by infection or cancer.