
Congo red, Ultra Pure Grade
Catalogue # 103011-118
Fournisseur: Anaspec Inc
Numéro CAS:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Spécifications
- Size:1 g
- Cat. no.:103011-118
- Supplier No.:AS-83016
Spécifications
À propos de cet article
:
:696.67 g/mol
Indice Merck:12,02562
Fusion Pt:> 360 °C
L'EINECS:209-358-4
:GHS08
Température de conservation:Ambient
Numéro CAS:573-58-0
Formule:C₃₂H₂₂N₆Na₂O₆S₂
Numéro MDL:MFCD00004028