To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Antibody type:Primary
- Antigen name:Seminal Fluid
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human
- Size:2 ml
- Form:Lyophilized powder
- Storage buffer:pH: 7.3
- Storage temperature:Store lyophilized product at 2-8°C. Store reconstituted material no longer than 4 weeks at 2-8°C. For long-term storage, aliquot
and freeze reconstituted product at –20 °C or lower. - Concentration:Total Protein: 34.5 mg/mL
- Shipping temperature:4C
- Cat. no.:IC0855137
Product is the lyophilized powder of rabbit antiserum to human serum and buffer salts.
The antiserum is prepared by hyperimmunizing rabbits with human serum. The antiserum is delipidated and dialyzed into 0.02M
sodium phosphate, 0.14M sodium chloride, pH 7.3, with 0.05% sodium azide, adjusted to standard titer, filtered through a 0.22
µm filter, vialed and lyophilized.
Type: Primary
Antigen: Seminal Fluid
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human