Order Entry
Canada
ContactUsLinkComponent
Gelatin, FITC (Fluorescein Isothiocyanate)
Catalog # 103011-298
Supplier:  Anaspec Inc
CAS Number:  
Gelatin, FITC (Fluorescein Isothiocyanate)
Catalog # 103011-298
Supplier:  Anaspec Inc
Supplier Number:  AS-85145
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Size:
    5 mg
  • Storage conditions:
    –20 °C desiccated and protected from light
  • Protein synonyms:
    Gelatine
  • Protein/peptide name:
    Gelatin
  • Formulation:
    in water
  • Shipping temperature:
    Shipped in Blue Ice
  • Cat. no.:
    103011-298
  • Supplier No.:
    AS-85145

Specifications

About this item

This gelatin is heavily labeled with FITC, and the conjugate is a highly quenched gelatinase/collagenase substrate. It is efficiently digested by gelatinases and collagenases, releasing brightly fluorescent peptides. The increase in fluorescence upon digestion is proportional to proteolytic activity. Longer incubation may increase its sensitivity for detecting proteases.

  • Absorbance: Abs/Em = 492/515 nm