Finger Goniometer, Sklar®
Finger Goniometer, Sklar®
Finger Goniometer, Stainless steel, 6"
Catalog # 76191-588
Supplier:   Sklar
Supplier #:  06-2820
Finger Goniometer, Sklar®
Catalog # 76191-588
Supplier:  Sklar
Finger Goniometer, Sklar®
Finger Goniometer, Sklar®
Finger Goniometer, Stainless steel, 6"
Price
$460.96
New Customer Promotional Price
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
All Sklar products are industry standard.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
Sklar's® Finger Goniometer is used to measure range of motion in fingers and other small joints like toes. The gauge measures a range of 30 to 120°. The instrument is stainless steel and is 6" overall length.
Specifications
  • Désignation:
    Finger Goniometer
  • Longueur:
    6"
  • Matière:
    Stainless steel