Aprotinin | Avantor
Aprotinin
100 mg
Catalog # CA82022-378
Supplier:   G-Biosciences
Fournisseur #:  786-046
Aprotinin
Catalog # CA82022-378
Supplier:  G-Biosciences
CAS Number:  
Aprotinin
Aprotinin
100 mg
CAS Number:  
Price
$2,096.03
Log in pour la disponibilité
Each (100mg)
La quantité minimale de commande est de 1
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents
A selection of individual protease inhibitors are offered for researchers who wish to design their own cocktails, supplement existing cocktails, or screen for specific proteases
  • Inhibits serine proteases (plasmin, kalikrein, trypsin and chymotrypsin).
Also available is the Protease Inhibitor Set, which contains all 12 of our individual protease inhibitors, each in a ready-to-use solution at a 100X concentration. The 1X concentration of the protease inhibitors is designed to give >90% inhibition in crude tissue extracts. Various concentrations and/or combinations of protease inhibitors may be used to inhibit a broad spectrum of protease activity.
G-Biosciences offers the following individual protease inhibitors as a dry powder: AEBSF, ALLN, Antipain dihydrochloride, Aprotinin, Bestatin, Chymostatin, E-64, EDTA-Na2, Leupeptin, Pepstatin, Phosphoramidon, and PMSF.
Spécifications
Spécifications
  • Taille:
    100 mg
  • Cat. No.:
    CA82022-378
  • Protein/peptide name:
    Aprotinin
Spécifications