Erlenmeyer Flasks, GL Thread, Chemglass
Erlenmeyer Flasks, GL Thread, Chemglass
Supplier:   Chemglass
Erlenmeyer Flasks, GL Thread, Chemglass
Supplier:  Chemglass
Certificates
Erlenmeyer Flasks, GL Thread, Chemglass
Erlenmeyer Flasks, GL Thread, Chemglass
6 Options Available Below
Product Details & Documents
These flasks are made of borosilicate glass.
  • Graduated to approximate volumes
  • With GL screw thread top
Delivery information: Supplied with polypropylene linerless closure and pouring ring.
Product Family Options
Product Information
  • Désignation
    Capacité
    Epaisseur
    Availability
    Price
  • Erlenmeyer Flask, GL Screw Thread
    125 ml
    GL 32
    Log in for availability
    Log in for availability
    Each
    Each - $112.00
    $112.00
    Each
       $112.00
    Minimum order quantity is 1
    Supplier Number: CG-1544-19
  • Erlenmeyer Flask, GL Screw Thread
    250 ml
    GL 45
    Log in for availability
    Log in for availability
    Each (250ml)
    Each (250ml) - $112.00
    $112.00
    Each (250ml)
       $112.00
    Minimum order quantity is 1
    Supplier Number: CG-1544-20
  • Erlenmeyer Flask, GL Screw Thread
    500 ml
    GL 45
    Log in for availability
    Log in for availability
    Each (500ml)
    Each (500ml) - $123.00
    $123.00
    Each (500ml)
       $123.00
    Minimum order quantity is 1
    Supplier Number: CG-1544-21
  • Erlenmeyer Flask, GL Screw Thread
    1000 ml
    GL 45
    Log in for availability
    Log in for availability
    Each (1l)
    Each (1l) - $151.00
    $151.00
    Each (1l)
       $151.00
    Minimum order quantity is 1
    Supplier Number: CG-1544-22
  • Erlenmeyer Flask, GL Screw Thread
    2000 ml
    GL 45
    Log in for availability
    Log in for availability
    Each (2l)
    Each (2l) - $205.00
    $205.00
    Each (2l)
       $205.00
    Minimum order quantity is 1
    Supplier Number: CG-1544-23
  • Erlenmeyer Flask, GL Screw Thread
    4000 ml
    GL 45
    Log in for availability
    Log in for availability
    Each (4l)
    Each (4l) - $386.00
    $386.00
    Each (4l)
       $386.00
    Minimum order quantity is 1
    Supplier Number: CG-1544-24