About this item
Endosafe®- PTS Endotoxin test utilizes existing FDA-licensed LAL formulations. The PTS provides quantitative LAL test results in approximately 15 minutes.
- Reference package insert for storage requirements
- Inhibition/enhancement screening
- Rapid Micro Methods – Glucan
- Acceptable temperature: 37 ±1 °C
- Sample requirement: 25 μl sample per channel
Rapid micro methods Gram ID: Archived parameters for gram-negative, gram positive and yeast/mold isolate identification.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.