SVL® Solid Caps, With PTFE Protected Seals, Chemglass
Fournisseur: Chemglass
Certificats
À propos de cet article
Solid SVL® caps with PTFE faced liner.
- Cap and liner are autoclavable upto 121 °C
These caps are compatible for use with CG-187 screw threads.
Frequently Bought Together