ContactUsLinkComponent
VWR® Economy Digital Thermometer
VWR® Economy Digital Thermometer
Catalog # 89140-510
Supplier:  VWR International
undefined
VWR® Economy Digital Thermometer
Catalog # 89140-510
Supplier:  VWR International
Specifications
  • Resolution:
    0.1
  • Description:
    VWR® Digital Thermometer with Probe
  • Environmentally Preferable:
  • Cat. no.:
    89140-510
  • Probe:
    Sensor with 39" immmersible cable
  • Range:
    –10...50 °C (14...122 °F), ambient; –50...70 °C (–58...158 °F), probe
Specifications
About this item
Economy digital thermometer with probe and used in ambient temperature.
  • High or low alarm throughput
  • With minimum and maximum memory
  • Available in °C and °F