Cole-Parmer® Insulated Cryogenic Gloves
Fournisseur: SPEX
Certificats
À propos de cet article
Cryogenic gloves designed to protect hands and arms from ultra-cold materials and liquid nitrogen exposure.
- Multi-layer insulation for optimal protection
- Flexible and lightweight to allow excellent dexterity
- Comfortable to wear for extended periods of time
Highly recommended for the safe operation of the Freezer/Mill®.
Frequently Bought Together