SureVector, Vectors, Agilent Technologies | Avantor
SureVector, Vectors, Agilent Technologies
SureVector, Vectors, Agilent Technologies
Certificates
SureVector, Vectors, Agilent Technologies
SureVector, Vectors, Agilent Technologies
3 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents
Combine standard DNA components to build custom vector constructs from Agilent’s set of standard parts.
  • Rapid custom vector generation - Less than a day from design to vector, compared to 4 weeks for custom vector services
  • Reliable and precise assembly - SureVector is extensively validated to ensure that standard parts can be interchanged, without loss of functionality
  • More flexible than traditional systems - Assemble new vectors in your lab as experimental requirements change, versus always ordering a new one
  • Control your experiments - Take control of your experiments, troubleshoot your own work, not that of your service providers DNA assembly
SureVector next-generation cloning uses the power of synthetic biology to combine standard DNA components to build custom vector constructs from the Agilent set of standard parts. Simply design a vector, order or PCR your gene-of-interest, and your custom plasmid is just a single reaction away.
Proprietary enzymes enable rapid and reliable assembly of multiple DNA fragments into a circular, recombinant plasmid containing your target gene.
Delivery information: Components include: bacterial selection markers, bacterial origins of replication, mammalian selection markers, promoters for yeast, mammalian cells and bacteria, and a variety of solubility/affinity tags; Enabling functionality in a range of cells.
Caution: For research use only. Not for use in diagnostic procedures.
Product Family Options
Informations sur le produit
  • Désignation
    Taille
    Details
    Availability
    Price
  • SureVector E. coli bacterial selection kit
    5 reactions
    SureVector E. coli bacterial selection kit. E. coli starter kit containing 3 separate bacterial selection markers
    Discontinued
    Supplier Number: -
    L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • SureVector E. coli N-terminal promoter kit
    5 reactions
    SureVector E. coli N-terminal promoter Kit. E. coli starter kit containing 3 separate promoters and a N-terminal His Tag
    Discontinued
    Supplier Number: -
    L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • SureVector E. coli C-terminal promoter kit
    5 reactions
    SureVector E. coli C-terminal promoter kit. E. coli starter kit containing 3 separate promoters and a C-terminal His Tag
    Discontinued
    Supplier Number: -
    L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • SureVector E. coli N-terminal tag kit
    5 reactions
    SureVector E. coli N-terminal tag Kit E. coli starter kit containing 3 separate N-terminal tags and a T7 promoter
    Discontinued
    Supplier Number: -
    L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • SureVector E. coli C-terminal tag kit
    5 reactions
    SureVector E. coli C-terminal tag kit. E. coli starter kit containing 3 separate C-terminal Tags and a T7 promoter
    Discontinued
    Supplier Number: -
    L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • SureVector Core kit
    15 reactions
    SureVector Core contains the basic elements needed for functionality in bacterial, yeast, and mammalian systems
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $676.24
    $676.24
    Each
       $676.24
    Supplier Number: G7514A
    L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • SureVector E. coli N-terminal expansion kit
    15 reactions
    SureVector E. coli N-terminal Expansion Kit expands functionality of the core kit in E. coli by including additional promoters and N-terminal tags
    Discontinued
    Supplier Number: -
    L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • SureVector E. coli C-terminal expansion kit
    15 reactions
    SureVector E. coli C-terminal Expansion Kit expands functionality of the core kit in E. coli by including additional promoters and C-terminal tags
    Discontinued
    Supplier Number: -
    L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • SureVector Mammalian N-terminal expansion kit
    15 reactions
    Promoters and N-terminal tags for expression in mammalian cells
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $949.00
    $949.00
    Each
       $949.00
    Supplier Number: G7516A
    L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • SureVector Mammalian C-terminal expansion kit
    15 reactions
    Promoters and C-terminal tags for expression in mammalian cells
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $949.00
    $949.00
    Each
       $949.00
    Supplier Number: G7516B
    L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • SureVector Yeast N-terminal expansion kit
    15 reactions
    Promoters and N-terminal tags for expression in yeast cells
    Discontinued
    Supplier Number: -
    L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • SureVector Yeast C-terminal expension kit
    15 reactions
    Promoters and C-terminal tags for expression in yeast cells
    Discontinued
    Supplier Number: -
    L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.