Brilliant II Probe MM with ROX, Agilent Technologies
Brilliant II QPCR and QRT-PCR reagents were developed to maintain the high quality of original Brilliant product line, but with a significant improvement in sensitivity of detection, ensuring reproducible quantification even at low target concentrations.
- Formulations available with low and high concentrations of ROX reference dye
- Improved performance with earlier Ct detection across a wide dynamic range
- Increased sensitivity of detection to low copy numbers
With the addition of ROX reference dye to our master mix, the reaction setup process is streamlined.
: For research use only. Not for use in diagnostic procedures.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Application:RT-qPCR probe
- One step:Yes
Frequently Bought Together