The AAV Helper-Free System eliminates the requirement for wild-type adenovirus co-infection from both AAV vector production and AAV stock titering steps, making this system entirely helper virus-free and safer. This system works with a broad range of hosts and infects both dividing and non-dividing cells.
- Stable gene expression
- High titer virus
- Lower toxicity vs. non-viral methods of gene delivery
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.